siRNA Details
| Gene Symbol: LOC101928259 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110391.1 | 1 | 32 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000420998.1 | - | 1 | 31 | Chr. X - 72688950 - 72712348 [+] on Build GRCh38 | |

LOC101928259
Genomic Map