siRNA Details
| Gene Symbol: LOC102724323 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120674.1 | 3 | 369 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000435635.1 | - | 3 | 369 | Chr. 10 - 45444570 - 45453121 [-] on Build GRCh38 | |

LOC102724323
Genomic Map