siRNA Details
| Gene Symbol: FLJ27354 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033981.1 | 2 | 279 | |||
| GenBank mRNA | AK130864.1 | - | 2 | 279 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FLJ27354| Gene Symbol: FLJ27354 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033981.1 | 2 | 279 | |||
| GenBank mRNA | AK130864.1 | - | 2 | 279 | ||