siRNA Details
| Gene Symbol: LOC101928475 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135638.1 | 2 | 162 | |||
| GenBank mRNA | BC033345.1 | - | 2 | 162 | ||
| BG722758.1 | - | 2 | 167 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000446671.1 | - | 2 | 162 | Chr. 17 - 15014805 - 15052276 [+] on Build GRCh38 | |

LOC101928475
Genomic Map