siRNA Details
| Gene Symbol: LINC01104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103730.1 | 3 | 1839 | |||
| GenBank mRNA | AK056640.1 | - | 3 | 1839 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000452354.5 | - | 3 | 1839 | Chr. 2 - 100208254 - 100251484 [+] on Build GRCh38 | |

LINC01104
Genomic Map