siRNA Details
Gene Symbol: LINC01104 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103730.1 | 1 | 193 | |||
GenBank mRNA | AK056640.1 | - | 1 | 193 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000452354.5 | - | 1 | 193 | Chr. 2 - 100208254 - 100251484 [+] on Build GRCh38 |
