siRNA Details
| Gene Symbol: LOC101929631 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110794.1 | 4 | 1580 | |||
| GenBank mRNA | BC039113.1 | - | 4 | 1584 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000456026.1 | - | 4 | 1580 | Chr. 1 - 218043505 - 218059140 [+] on Build GRCh38 | |

LOC101929631
Genomic Map