siRNA Details
| Gene Symbol: LINC00867 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108045.1 | 2, 3 | 380 | |||
| GenBank mRNA | AI733769.1 | - | 2, 3 | 382 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00867| Gene Symbol: LINC00867 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108045.1 | 2, 3 | 380 | |||
| GenBank mRNA | AI733769.1 | - | 2, 3 | 382 | ||