siRNA Details
Gene Symbol: RAP2C-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110410.1 | 3 | 2228 | |||
GenBank mRNA | AK095439.1 | - | 3 | 2114 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000441399.2 | - | 3 | 2228 | Chr. X - 132218507 - 132432862 [+] on Build GRCh38 |
