siRNA Details
| Gene Symbol: RAP2C-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110410.1 | 3 | 3303 | |||
| GenBank mRNA | AK095439.1 | - | 3 | 3189 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000441399.2 | - | 3 | 3303 | Chr. X - 132218507 - 132432862 [+] on Build GRCh38 | |

RAP2C-AS1
Genomic Map