siRNA Details
| Gene Symbol: LINC00708 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108058.1 | 4 | 560 | |||
| GenBank mRNA | BC031880.1 | - | 4 | 571 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00708| Gene Symbol: LINC00708 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108058.1 | 4 | 560 | |||
| GenBank mRNA | BC031880.1 | - | 4 | 571 | ||