siRNA Details
| Gene Symbol: LOC440390 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126008.1 | 3 | 3044 | |||
| GenBank mRNA | AK128193.1 | - | 3 | 3044 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000430050.2 | - | 3 | 3044 | Chr. 16 - 87057784 - 87063991 [+] on Build GRCh38 | |

LOC440390
Genomic Map