siRNA Details
| Gene Symbol: LINC00202-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026794.1 | 1 | 4157 | |||
| GenBank mRNA | CR749234.1 | - | 1 | 4097 | ||
| Gene Symbol: LINC00202-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK097405.1 | - | 1 | 114 | ||
| AK128027.1 | - | 1 | 4946 | |||

LINC00202-2
Genomic Map