siRNA Details
| Gene Symbol: LINC00865 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038382.1 | 4 | 2483 | |||
| GenBank mRNA | AL137595.1 | - | 3 | 2085 | ||
| BC033541.1 | - | 4 | 2466 | |||
| BC035398.1 | - | 4 | 2466 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00865| Gene Symbol: LINC00865 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038382.1 | 4 | 2483 | |||
| GenBank mRNA | AL137595.1 | - | 3 | 2085 | ||
| BC033541.1 | - | 4 | 2466 | |||
| BC035398.1 | - | 4 | 2466 | |||