siRNA Details
Gene Symbol: LOC101928304 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125855.1 | 2 | 295 | |||
NR_125856.1 | 2 | 295 | ||||
GenBank mRNA | AV705921.2 | - | 2 | 328 | ||
DA256978.1 | - | 2 | 282 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101928304 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125855.1 | 2 | 295 | |||
NR_125856.1 | 2 | 295 | ||||
GenBank mRNA | AV705921.2 | - | 2 | 328 | ||
DA256978.1 | - | 2 | 282 |