siRNA Details
| Gene Symbol: LINC00112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024028.2 | 1 | 17 | |||
| GenBank mRNA | AY040089.1 | - | 1 | 17 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000432830.1 | - | 1 | 17 | Chr. 21 - 41716436 - 41717580 [+] on Build GRCh38 | |

LINC00112
Genomic Map