siRNA Details
| Gene Symbol: LOC100505771 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132417.1 | 2 | 564 | |||
| NR_132418.1 | 2 | 485 | ||||
| GenBank mRNA | BM263220.1 | - | 2 | 470 | ||
| BQ220589.1 | - | 2 | 564 | |||
| BU500705.1 | - | 1 | 126 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100505771| Gene Symbol: LOC100505771 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132417.1 | 2 | 564 | |||
| NR_132418.1 | 2 | 485 | ||||
| GenBank mRNA | BM263220.1 | - | 2 | 470 | ||
| BQ220589.1 | - | 2 | 564 | |||
| BU500705.1 | - | 1 | 126 | |||