siRNA Details
| Gene Symbol: LOC151484 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038238.1 | 3 | 1149 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC151484| Gene Symbol: LOC151484 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038238.1 | 3 | 1149 | |||