siRNA Details
| Gene Symbol: LOC100507507 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001322263.1 | NP_001309192.1 | 2 | 893 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000429630.1 | - | 2 | 363 | Chr. 7 - 143379692 - 143380495 [-] on Build GRCh38 | |

LOC100507507
Genomic Map