siRNA Details
| Gene Symbol: LOC101927257 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109965.1 | 1 | 426 | |||
| GenBank mRNA | AK097769.1 | - | 1 | 426 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000417123.1 | - | 1 | 426 | Chr. 22 - 40521800 - 40526707 [+] on Build GRCh38 | |

LOC101927257
Genomic Map