siRNA Details
Gene Symbol: LOC101927143 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110669.1 | 2 | 770 | |||
GenBank mRNA | BC008027.1 | - | 2 | 770 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000436706.1 | - | 2 | 770 | Chr. 1 - 224208747 - 224213279 [-] on Build GRCh38 |
