siRNA Details
| Gene Symbol: LINC01431 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109884.1 | 3 | 378 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000452395.1 | - | 2 | 448 | Chr. 20 - 23356795 - 23358116 [-] on Build GRCh38 | |

LINC01431
Genomic Map