siRNA Details
Gene Symbol: LOC100507670 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135595.1 | 2 | 538 | |||
GenBank mRNA | BC021024.1 | - | 2 | 538 | ||
BE385446.1 | - | 2 | 566 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000447795.3 | - | 2 | 538 | Chr. 1 - 151280024 - 151281929 [-] on Build GRCh38 |
