siRNA Details
| Gene Symbol: LINC01628 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110600.1 | 3 | 307 | |||
| GenBank mRNA | BX103810.1 | - | 3 | 307 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000444266.1 | - | 3 | 274 | Chr. 2 - 66921510 - 66922457 [-] on Build GRCh38 | |

LINC01628
Genomic Map