siRNA Details
| Gene Symbol: KIZ-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109956.1 | 2 | 196 | |||
| GenBank mRNA | BC034426.1 | - | 2 | 195 | ||
| BC042893.1 | - | 2 | 172 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KIZ-AS1| Gene Symbol: KIZ-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109956.1 | 2 | 196 | |||
| GenBank mRNA | BC034426.1 | - | 2 | 195 | ||
| BC042893.1 | - | 2 | 172 | |||