siRNA Details
| Gene Symbol: LOC101926943 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110044.1 | 1, 2 | 209 | |||
| GenBank mRNA | DA836308.1 | - | 1, 2 | 209 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101926943| Gene Symbol: LOC101926943 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110044.1 | 1, 2 | 209 | |||
| GenBank mRNA | DA836308.1 | - | 1, 2 | 209 | ||