siRNA Details
| Gene Symbol: LOC105376955 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136189.1 | 3 | 552 | |||
| GenBank mRNA | BG759368.1 | - | 3 | 551 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000448129.1 | - | 3 | 552 | Chr. 3 - 12877522 - 12885211 [-] on Build GRCh38 | |

LOC105376955
Genomic Map