siRNA Details
| Gene Symbol: LOC101928435 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109962.1 | 2 | 82 | |||
| GenBank mRNA | BC034606.1 | - | 2 | 82 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928435| Gene Symbol: LOC101928435 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109962.1 | 2 | 82 | |||
| GenBank mRNA | BC034606.1 | - | 2 | 82 | ||