siRNA Details
| Gene Symbol: LOC101928435 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109962.1 | 3 | 1001 | |||
| GenBank mRNA | BC034606.1 | - | 3 | 1001 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000433952.1 | - | 3 | 1001 | Chr. 21 - 38974429 - 38977774 [-] on Build GRCh38 | |

LOC101928435
Genomic Map