siRNA Details
| Gene Symbol: LOC101929413 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109866.1 | 7 | 1127 | |||
| GenBank mRNA | BC033557.1 | - | 7 | 1127 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000448859.1 | - | 7 | 1127 | Chr. 20 - 10875333 - 10909272 [-] on Build GRCh38 | |

LOC101929413
Genomic Map