siRNA Details
Gene Symbol: LINC01166 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120626.1 | 2 | 2659 | |||
NR_120627.1 | 2 | 2493 | ||||
GenBank mRNA | AK095503.1 | - | 2 | 2493 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000443633.1 | - | 2 | 2493 | Chr. 10 - 132943967 - 132965424 [-] on Build GRCh38 |
