siRNA Details
| Gene Symbol: LINC01166 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120626.1 | 2 | 2686 | |||
| NR_120627.1 | 2 | 2520 | ||||
| GenBank mRNA | AK095503.1 | - | 2 | 2520 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000443633.1 | - | 2 | 2520 | Chr. 10 - 132943967 - 132965424 [-] on Build GRCh38 | |

LINC01166
Genomic Map