siRNA Details
Gene Symbol: DLGAP4-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109939.1 | 5 | 1037 | |||
GenBank mRNA | BC039668.1 | - | 5 | 1037 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000439595.5 | - | 1037 | Chr. 20 - 36508160 - 36573275 [-] on Build GRCh38 |
