siRNA Details
Gene Symbol: PLCE1-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120615.1 | 3 | 1115 | |||
GenBank mRNA | BC035380.1 | - | 3 | 1106 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000432782.1 | - | 3 | 1106 | Chr. 10 - 94099648 - 94108785 [-] on Build GRCh38 |
