siRNA Details
| Gene Symbol: PLCE1-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120615.1 | 1 | 336 | |||
| GenBank mRNA | BC035380.1 | - | 1 | 327 | ||
| HY019250.1 | - | 1 | 337 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PLCE1-AS2| Gene Symbol: PLCE1-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120615.1 | 1 | 336 | |||
| GenBank mRNA | BC035380.1 | - | 1 | 327 | ||
| HY019250.1 | - | 1 | 337 | |||