siRNA Details
| Gene Symbol: LINC00342 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK057592.1 | - | 1 | 963 | ||
| AK057596.1 | - | 1 | 1640 | |||
| AK093195.1 | - | 1 | 456 | |||
| BC117383.1 | - | 1 | 1606 | |||
| BC126153.1 | - | 1 | 1603 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000448494.1 | - | 1 | 1088 | Chr. 2 - 95807118 - 95816215 [-] on Build GRCh38 | |

LINC00342
Genomic Map