siRNA Details
| Gene Symbol: RGS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045630.1 | 4 | 1101 | |||
| GenBank mRNA | BC032239.1 | - | 4 | 1062 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000439699.1 | - | 4 | 1062 | Chr. 1 - 163304211 - 163321752 [-] on Build GRCh38 | |

RGS5
Genomic Map