siRNA Details
| Gene Symbol: LOC101927560 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_119374.1 | 3 | 1361 | |||
| GenBank mRNA | AK097722.1 | - | 3 | 1361 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000457273.5 | - | 3 | 1361 | Chr. 1 - 83801516 - 83860546 [-] on Build GRCh38 | |

LOC101927560
Genomic Map