siRNA Details
| Gene Symbol: LOC101929486 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109868.1 | 1 | 537 | |||
| GenBank mRNA | BC036700.1 | - | 1 | 542 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000456265.1 | - | 1 | 537 | Chr. 20 - 12865202 - 12952519 [-] on Build GRCh38 | |

LOC101929486
Genomic Map