siRNA Details
Gene Symbol: ERVH48-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308491.1 | NP_001295420.1 | 2 | 1906 | ||
GenBank mRNA | AB610407.1 | - | 2 | 1906 | ||
BC005107.1 | - | 2 | 1926 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000447535.1 | - | 2 | 1954 | Chr. 21 - 42916803 - 42925646 [-] on Build GRCh38 |
