siRNA Details
| Gene Symbol: ERVH48-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308491.1 | NP_001295420.1 | 2 | 1595 | ||
| GenBank mRNA | AB610407.1 | - | 2 | 1595 | ||
| BC005107.1 | - | 2 | 1615 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000447535.1 | - | 2 | 1643 | Chr. 21 - 42916803 - 42925646 [-] on Build GRCh38 | |

ERVH48-1
Genomic Map