siRNA Details
| Gene Symbol: LINC01271 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109950.1 | 2 | 285 | |||
| GenBank mRNA | BC039367.1 | - | 2 | 285 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000441214.1 | - | 2 | 285 | Chr. 20 - 50310711 - 50321342 [-] on Build GRCh38 | |

LINC01271
Genomic Map