siRNA Details
Gene Symbol: LINC01271 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109950.1 | 3 | 1980 | |||
GenBank mRNA | BC039367.1 | - | 3 | 1980 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000441214.1 | - | 3 | 1980 | Chr. 20 - 50310711 - 50321342 [-] on Build GRCh38 |
