siRNA Details
| Gene Symbol: LINC00892 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC042469.1 | - | 2 | 190 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000427517.1 | - | 2 | 190 | Chr. X - 136639740 - 136642429 [+] on Build GRCh38 | |

LINC00892
Genomic Map