siRNA Details
| Gene Symbol: LINC00892 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038461.1 | 3 | 588 | |||
| NR_038462.1 | 3 | 555 | ||||
| GenBank mRNA | BC042469.1 | - | 2 | 1619 | ||
| BG189095.1 | - | 1 | 234 | |||
| BX112139.1 | - | 3 | 358 | |||
| CD699893.1 | - | 3 | 452 | |||

LINC00892
Genomic Map