siRNA Details
Gene Symbol: LINC00702 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108040.1 | 2 | 189 | |||
GenBank mRNA | DA853538.1 | - | 2 | 189 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000454470.1 | - | 1 | 55 | Chr. 10 - 4235560 - 4243643 [-] on Build GRCh38 |
