siRNA Details
| Gene Symbol: LINC01007 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103747.1 | 2 | 2239 | |||
| NR_103748.1 | 2 | 1392 | ||||
| GenBank mRNA | AF131787.1 | - | 2 | 1392 | ||
| BC027906.1 | - | 2 | 2183 | |||
| BC039094.1 | - | 2 | 1341 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01007| Gene Symbol: LINC01007 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103747.1 | 2 | 2239 | |||
| NR_103748.1 | 2 | 1392 | ||||
| GenBank mRNA | AF131787.1 | - | 2 | 1392 | ||
| BC027906.1 | - | 2 | 2183 | |||
| BC039094.1 | - | 2 | 1341 | |||