siRNA Details
Gene Symbol: UBE2E2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136176.1 | 2 | 305 | |||
NR_136177.1 | 2 | 329 | ||||
GenBank mRNA | HY029877.1 | - | 2 | 308 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: UBE2E2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136176.1 | 2 | 305 | |||
NR_136177.1 | 2 | 329 | ||||
GenBank mRNA | HY029877.1 | - | 2 | 308 |