siRNA Details
| Gene Symbol: LOC102606465 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109849.1 | 5 | 1063 | |||
| GenBank mRNA | AK021551.1 | - | 2 | 1386 | ||
| BC045807.2 | - | 7 | 1147 | |||
| BX538249.1 | - | 9 | 1875 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102606465| Gene Symbol: LOC102606465 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109849.1 | 5 | 1063 | |||
| GenBank mRNA | AK021551.1 | - | 2 | 1386 | ||
| BC045807.2 | - | 7 | 1147 | |||
| BX538249.1 | - | 9 | 1875 | |||