siRNA Details
| Gene Symbol: LOC100996842 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136401.1 | 1 | 99 | |||
| NR_136402.1 | 1 | 99 | ||||
| GenBank mRNA | BM888151.1 | - | 1 | 88 | ||
| HY076064.1 | - | 1 | 100 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000417897.1 | - | 1 | 88 | Chr. 17 - 7582016 - 7584068 [-] on Build GRCh38 | |

LOC100996842
Genomic Map